Anti-SNRPD2P2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-70708
102200-014EA 536.43 USD
102200-014
Anti-SNRPD2P2 Rabbit Polyclonal Antibody
Antibodies
The SNRPD2P2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SNRPD2P2. This antibody reacts with human. The SNRPD2P2 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SNRPD2P2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR