Anti-SNRPC Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-57153
102189-206EA 564.74 USD
102189-206
Anti-SNRPC Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal SNRPC Antibody. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human.

Type: Primary
Antigen: SNRPC
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR