Anti-SNRP70 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-57488
102189-864EA 564.74 USD
102189-864
Anti-SNRP70 Rabbit Polyclonal Antibody
Antibodies
The SNRP70 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SNRP70. This antibody reacts with human. The SNRP70 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SNRP70
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR