Anti-SnoN Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-77306
102211-398EA 564.74 USD
102211-398
Anti-SnoN Rabbit Polyclonal Antibody
Antibodies
The SnoN Antibody from Novus Biologicals is a rabbit polyclonal antibody to SnoN. This antibody reacts with human, mouse, rat. The SnoN Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.

Type: Primary
Antigen: SnoN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR