Anti-SNIP1 Rabbit Polyclonal Antibody

NB100-512
102109-426EA 536.92 USD
102109-426
Anti-SNIP1 Rabbit Polyclonal Antibody
The SNIP1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SNIP1. This antibody reacts with human. The SNIP1 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.

Type: Primary
Antigen: SNIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR