Anti-SNF8 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-03503
102156-008EA 299 USD
102156-008
Anti-SNF8 Rabbit Polyclonal Antibody
Antibodies
The SNF8 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SNF8. This antibody reacts with human, mouse, rat, xenopus. The SNF8 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Recommended Dilutions: Immunohistochemistry (Parafin): 10-50 ?g/ml

Type: Primary
Antigen: SNF8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Xenopus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR