Anti-SNAP29 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-79769
102215-186EA 564.74 USD
102215-186
Anti-SNAP29 Rabbit Polyclonal Antibody
Antibodies
The SNAP29 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SNAP29. This antibody reacts with rat. The SNAP29 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SNAP29
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR