Anti-SNAP25 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-33714
102167-360EA 635.51 USD
102167-360
Anti-SNAP25 Rabbit Polyclonal Antibody
Antibodies
The SNAP25 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SNAP25. This antibody reacts with human, mouse, rat. The SNAP25 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100-1:500

Type: Primary
Antigen: SNAP25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR