Anti-SNAP alpha Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-79955
102215-558EA 299 USD
102215-558
Anti-SNAP alpha Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal SNAP alpha Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse, Rat, Bovine, Canine, Xenopus, Zebrafish.

Type: Primary
Antigen: SNAP alpha
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR