Anti-smu1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-56881
102188-678EA 564.74 USD
102188-678
Anti-smu1 Rabbit Polyclonal Antibody
Antibodies
The smu1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to smu1. This antibody reacts with human. The smu1 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: smu1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR