Anti-SMR3A Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-58000
102190-698EA 564.74 USD
102190-698
Anti-SMR3A Rabbit Polyclonal Antibody
Antibodies
The SMR3A Antibody from Novus Biologicals is a rabbit polyclonal antibody to SMR3A. This antibody reacts with human. The SMR3A Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SMR3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR