Anti-SMOC-2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-39788
102168-958EA 635.51 USD
102168-958
Anti-SMOC-2 Rabbit Polyclonal Antibody
Antibodies
The SMOC-2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SMOC-2. This antibody reacts with human. The SMOC-2 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 20 ?g/ml

Type: Primary
Antigen: SMOC-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR