Anti-SMG7 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-22967
102160-772EA 635.51 USD
102160-772
Anti-SMG7 Rabbit Polyclonal Antibody
Antibodies
The SMG7 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SMG7. This antibody reacts with human, mouse. The SMG7 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.

Type: Primary
Antigen: SMG7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR