Anti-SMC3 Rabbit Polyclonal Antibody (Agarose Immobilized)

NB600-434
102133-968EA 635.38 USD
102133-968
Anti-SMC3 Rabbit Polyclonal Antibody (Agarose Immobilized)
The SMC3 Antibody [Agarose Immobilized] from Novus Biologicals is a rabbit polyclonal antibody to SMC3. This antibody reacts with human, mouse. The SMC3 Antibody [Agarose Immobilized] has been validated for the following applications: Immunoprecipitation.

Type: Primary
Antigen: SMC3
Clonality: Polyclonal
Clone:
Conjugation: Agarose Immobilized
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR