Anti-SMC1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-205
102106-558EA 706.28 USD
102106-558
Anti-SMC1 Rabbit Polyclonal Antibody
Antibodies
The SMC1 [p Ser957] Antibody from Novus Biologicals is a rabbit polyclonal antibody to SMC1. This antibody reacts with human. The SMC1 [p Ser957] Antibody has been validated for the following applications: Western Blot, Immunoprecipitation, Proximity Ligation Assay.

Type: Primary
Antigen: SMC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR