Anti-SMARCA6 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-278
102108-154EA 635.51 USD
102108-154
Anti-SMARCA6 Rabbit Polyclonal Antibody
Antibodies
The SMARCA6 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SMARCA6. This antibody reacts with human. The SMARCA6 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.

Type: Primary
Antigen: SMARCA6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR