Anti-Smad1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-77858
102212-136EA 777.05 USD
102212-136
Anti-Smad1 Rabbit Polyclonal Antibody
Antibodies
The Smad1 [p Ser206] Antibody from Novus Biologicals is a rabbit polyclonal antibody to Smad1. This antibody reacts with human, mouse, rat, canine. The Smad1 [p Ser206] Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: Smad1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Canine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR