Anti-Transgelin Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
30920002
102103-310EA 399 USD
102103-310
Anti-Transgelin Rabbit Polyclonal Antibody
Rabbit Polyclonal SM22 alpha Antibody. Tested Applications: Western Blot, ELISA. Tested Reactivity: Human.

Type: Primary Antibodies
Antigen: Transgelin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR