Anti-SLTM Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-47299
102177-012EA 635.51 USD
102177-012
Anti-SLTM Rabbit Polyclonal Antibody
Antibodies
The SLTM Antibody from Novus Biologicals is a rabbit polyclonal antibody to SLTM. This antibody reacts with human. The SLTM Antibody has been validated for the following applications: Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SLTM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR