Anti-SLITRK6 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-76876
102209-714EA 564.74 USD
102209-714
Anti-SLITRK6 Rabbit Polyclonal Antibody
Antibodies
The SLITRK6 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SLITRK6. This antibody reacts with human, mouse, rat. The SLITRK6 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SLITRK6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR