Anti-SLITRK4 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-76875
102209-710EA 564.74 USD
102209-710
Anti-SLITRK4 Rabbit Polyclonal Antibody
Antibodies
The SLITRK4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SLITRK4. This antibody reacts with human, mouse, rat. The SLITRK4 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.

Type: Primary
Antigen: SLITRK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR