Anti-SLITRK1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-77319
102211-450EA 564.74 USD
102211-450
Anti-SLITRK1 Rabbit Polyclonal Antibody
Antibodies
The SLITRK1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SLITRK1. This antibody reacts with human, mouse, rat. The SLITRK1 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.

Type: Primary
Antigen: SLITRK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR