Anti-SLITRK1 Rabbit Polyclonal Antibody (DyLight 488)

Supplier: Novus Biologicals
NBP1-49541G
102179-188EA 614.15 USD
102179-188
Anti-SLITRK1 Rabbit Polyclonal Antibody (DyLight 488)
Antibodies
The SLITRK1 Antibody [DyLight 488] from Novus Biologicals is a rabbit polyclonal antibody to SLITRK1. This antibody reacts with human, mouse. The SLITRK1 Antibody [DyLight 488] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: SLITRK1
Clonality: Polyclonal
Clone:
Conjugation: DyLight 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR