Anti-SLIRP Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Novus Biologicals
NB110-37258H
102121-764EA 614.15 USD
102121-764
Anti-SLIRP Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
The SLIRP Antibody [HRP] from Novus Biologicals is a rabbit polyclonal antibody to SLIRP. This antibody reacts with human, mouse, primate. The SLIRP Antibody [HRP] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SLIRP
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR