Anti-SLCO3A1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-59678
102193-044EA 564.74 USD
102193-044
Anti-SLCO3A1 Rabbit Polyclonal Antibody
Antibodies
The SLCO3A1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SLCO3A1. This antibody reacts with human. The SLCO3A1 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SLCO3A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR