Anti-SLC9A4 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-46583
102176-398EA 564.74 USD
102176-398
Anti-SLC9A4 Rabbit Polyclonal Antibody
Antibodies
The SLC9A4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SLC9A4. This antibody reacts with human, mouse, rat. The SLC9A4 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry.

Type: Primary
Antigen: SLC9A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR