Anti-CAT3/SLC7A3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-78911
102213-474EA 429 USD
102213-474
Anti-CAT3/SLC7A3 Rabbit Polyclonal Antibody
The CAT3 / SLC7A3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CAT3 / SLC7A3. This antibody reacts with human. The CAT3 / SLC7A3 Antibody has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:1000

Type: Primary
Antigen: CAT3/SLC7A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR