Anti-CAT3/SLC7A3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-74144
102205-030EA 564.74 USD
102205-030
Anti-CAT3/SLC7A3 Rabbit Polyclonal Antibody
Antibodies
The CAT3 / SLC7A3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CAT3 / SLC7A3. This antibody reacts with mouse. The CAT3 / SLC7A3 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: CAT3/SLC7A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR