Anti-SLC6A8 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-60082
102193-836EA 564.74 USD
102193-836
Anti-SLC6A8 Rabbit Polyclonal Antibody
Antibodies
The SLC6A8 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SLC6A8. This antibody reacts with human. The SLC6A8 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SLC6A8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR