Anti-SLC6A18 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-59903
102193-486EA 564.74 USD
102193-486
Anti-SLC6A18 Rabbit Polyclonal Antibody
Antibodies
The SLC6A18 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SLC6A18. This antibody reacts with human. The SLC6A18 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SLC6A18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR