Anti-SLC6A15 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-74198
102205-138EA 564.74 USD
102205-138
Anti-SLC6A15 Rabbit Polyclonal Antibody
Antibodies
The SLC6A15 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SLC6A15. This antibody reacts with mouse. The SLC6A15 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SLC6A15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR