Anti-SLC5A9 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-59873
102193-430EA 564.74 USD
102193-430
Anti-SLC5A9 Rabbit Polyclonal Antibody
Antibodies
The SLC5A9 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SLC5A9. This antibody reacts with human. The SLC5A9 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SLC5A9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR