Anti-SLC5A8 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-62527
102195-688EA 564.74 USD
102195-688
Anti-SLC5A8 Rabbit Polyclonal Antibody
Antibodies
The SLC5A8 / SMCT1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SLC5A8 / SMCT1. This antibody reacts with human. The SLC5A8 / SMCT1 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SLC5A8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR