Anti-SLC4A5 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-59528
102192-748EA 564.74 USD
102192-748
Anti-SLC4A5 Rabbit Polyclonal Antibody
Antibodies
The SLC4A5 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SLC4A5. This antibody reacts with human. The SLC4A5 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SLC4A5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR