Anti-SLC46A3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-80531
102216-708EA 564.74 USD
102216-708
Anti-SLC46A3 Rabbit Polyclonal Antibody
Antibodies
The SLC46A3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SLC46A3. This antibody reacts with human. The SLC46A3 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SLC46A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR