Anti-SLC45A2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-18781
102157-896EA 329 USD
102157-896
Anti-SLC45A2 Rabbit Polyclonal Antibody
Antibodies
The SLC45A2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SLC45A2. This antibody reacts with human, mouse. The SLC45A2 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:50 to 1:500

Type: Primary
Antigen: SLC45A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR