Anti-SLC43A3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-69535
102199-080EA 564.74 USD
102199-080
Anti-SLC43A3 Rabbit Polyclonal Antibody
Antibodies
The SLC43A3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SLC43A3. This antibody reacts with human. The SLC43A3 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SLC43A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR