Anti-SLC39A3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-76500
102208-260EA 564.74 USD
102208-260
Anti-SLC39A3 Rabbit Polyclonal Antibody
Antibodies
The SLC39A3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SLC39A3. This antibody reacts with human, mouse. The SLC39A3 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry.

Type: Primary
Antigen: SLC39A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR