Anti-SLC31A1/CTR1 Rabbit Polyclonal Antibody (DyLight 550)

Supplier: Novus Biologicals
NB100-402R
102108-828EA 614.15 USD
102108-828
Anti-SLC31A1/CTR1 Rabbit Polyclonal Antibody (DyLight 550)
Antibodies
The SLC31A1 / CTR1 Antibody [DyLight 550] from Novus Biologicals is a rabbit polyclonal antibody to SLC31A1 / CTR1. This antibody reacts with human, mouse, rat, porcine, xenopus, zebrafish. The SLC31A1 / CTR1 Antibody [DyLight 550] has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:250

Type: Primary
Antigen: SLC31A1/CTR1
Clonality: Polyclonal
Clone:
Conjugation: DyLight 550
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Porcine, Xenopus, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR