Anti-SLC2A11 Rabbit Polyclonal Antibody

NBP1-46401
102176-138EA 395.38 USD
102176-138
Anti-SLC2A11 Rabbit Polyclonal Antibody
The SLC2A11 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SLC2A11. This antibody reacts with human. The SLC2A11 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry.

Type: Primary
Antigen: SLC2A11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR