Anti-SLC26A1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-59652
102192-992EA 564.74 USD
102192-992
Anti-SLC26A1 Rabbit Polyclonal Antibody
Antibodies
The SLC26A1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SLC26A1. This antibody reacts with human. The SLC26A1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: SLC26A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR