Anti-SLC25A20 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-59574
102192-840EA 564.74 USD
102192-840
Anti-SLC25A20 Rabbit Polyclonal Antibody
Antibodies
The SLC25A20 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SLC25A20. This antibody reacts with human, rat. The SLC25A20 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SLC25A20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR