Anti-SLC22A7 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-69691
102199-300EA 564.74 USD
102199-300
Anti-SLC22A7 Rabbit Polyclonal Antibody
Antibodies
The SLC22A7 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SLC22A7. This antibody reacts with human. The SLC22A7 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SLC22A7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR