Anti-SLC22A6 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-69694
102199-304EA 564.74 USD
102199-304
Anti-SLC22A6 Rabbit Polyclonal Antibody
Antibodies
The SLC22A6 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SLC22A6. This antibody reacts with human. The SLC22A6 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SLC22A6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR