Anti-SLC22A10 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-91568
102217-674EA 564.74 USD
102217-674
Anti-SLC22A10 Rabbit Polyclonal Antibody
Antibodies
The SLC22A10 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SLC22A10. This antibody reacts with human. The SLC22A10 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SLC22A10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR