Anti-SLC12A8 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-59893
102193-466EA 564.74 USD
102193-466
Anti-SLC12A8 Rabbit Polyclonal Antibody
Antibodies
The SLC12A8 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SLC12A8. This antibody reacts with human. The SLC12A8 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SLC12A8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR