Anti-SLC12A5 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-74063
102204-870EA 564.74 USD
102204-870
Anti-SLC12A5 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal SLC12A5 Antibody. Tested Applications: Western Blot. Tested Reactivity: Mouse.

Type: Primary
Antigen: SLC12A5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR