Anti-SLC10A7 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-59875
102193-432EA 564.74 USD
102193-432
Anti-SLC10A7 Rabbit Polyclonal Antibody
Antibodies
The SLC10A7 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SLC10A7. This antibody reacts with human. The SLC10A7 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SLC10A7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR