Anti-SKI Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-77305
102211-394EA 564.74 USD
102211-394
Anti-SKI Rabbit Polyclonal Antibody
Antibodies
The SKI Antibody from Novus Biologicals is a rabbit polyclonal antibody to SKI. This antibody reacts with human. The SKI Antibody has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence, Immunofluorescence.

Type: Primary
Antigen: SKI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR