Anti-SKA3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-76311
102207-530EA 564.74 USD
102207-530
Anti-SKA3 Rabbit Polyclonal Antibody
Antibodies
The SKA3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SKA3. This antibody reacts with human, mouse, rat. The SKA3 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SKA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR